Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCSER2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557968 Supplier Invitrogen™ Supplier No. PA557968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVECDNMNRF DRPDRNVRQP QEGFWKRPPQ RWSGQEHYHL SHPDHYHHHG KSDLSRGSPY RESPLGHFES YGGMPFFQAQ KMFVDVPENT VILDEMT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCSER2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCSER2
Gene Accession No. Q3UHI0, Q9H7U1
Gene Alias 1700012P13Rik; 2900054P12Rik; AI035535; AW108503; bA486O22.1; Ccser2; coiled-coil serine rich 2; coiled-coil serine rich protein 2; coiled-coil serine-rich protein 2; FAM190B; family with sequence similarity 190, member B; Gcap14; granule cell antiserum positive 14; granule cell antiserum positive protein 14; KIAA1128; NPD012; protein FAM190B; Protein GCAP14; Protein GCAP14 homolog; RGD1304626; serine-rich coiled-coil domain-containing protein 2
Gene Symbols CCSER2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCSER2. Recombinant protein control fragment (Product #RP-97047).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306306, 54462, 72972
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.