Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CCT 018159
GREENER_CHOICE

Catalog No. 243510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-351-0 10 mg
24-355-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-351-0 Supplier Tocris Bioscience™ Supplier No. 2435/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hsp90 inhibitor

Novel inhibitor of heat shock protein 90 (Hsp90) ATPase activity (IC50 = 5.7 μM) that displays selectivity over human Hsp72 and topoisomerase II. Inhibits proliferation of HCT116 human colon tumor cells and produces upregulation of Hsp70 and downregulation of c-Raf and cdk4. More soluble than 17-AAG (Cat. No. 1515) and is independent of NQO1/DT-diaphorase and P-glycoprotein expression.

Chemical Identifiers

CAS 171009-07-7
Molecular Formula C20H20N2O4

Specifications

Quantity 10 mg
Formula Weight 352.39
Purity >98%
Molecular Weight (g/mol) 352.39
Chemical Name or Material CCT 018159
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.