Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117055 Supplier Invitrogen™ Supplier No. PA5117055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, 293T, HepG2, Mouse brain, Mouse heart, Mouse kidney, Mouse liver Immunogen sequence: MMGHRPVLVL SQNTKRESGR KVQSGNINAA KTIADIIRTC LGPKSMMKML LDPMGGIVMT NDGNAILREI QVQHPAAKSM IEISRTQDEE VGDGTTSVII LAGEMLSVAE HFLEQQMHPT VVISAYRKAL DDMISTLKKI SIPVDISDSD MMLNIINSSI TTKAISRWSS LACNIALDAV KMVQFEENGR KEIDIKKYAR VEKIPGGIIE DSCVLRGVMI NKDVTHPRMR RYIKNPRIVL LDSSLEYKKG ESQTDIEITR EEDFTRILQM EEEYIQQLCE DIIQLKPDVV ITEKGISDLA.

The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants have been characterized for this gene. In addition, a pseudogene of this gene has been found on chromosome 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCT3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 6.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cct3
Gene Accession No. P49368, P80318, Q6P502
Gene Alias AL024092; CCT3; CCTG; CCT-gamma; chaperonin containing TCP1 subunit 3; chaperonin containing Tcp1, subunit 3 (gamma); chaperonin subunit 3 (gamma); hTRiC5; Matricin; mTRiC-P5; OTTHUMP00000025735; PIG48; T-complex protein 1 subunit gamma; T-complex protein 1, gamma subunit; TCP1 (t-complex-1) ring complex, polypeptide 5; TCP-1-gamma; Tcp1-rs3; TRIC5; TriC-P5
Gene Symbols Cct3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CCT3 (NP_0059893).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12462, 295230, 7203
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.