Learn More
Invitrogen™ CCT3 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human A549 whole cell, human Hela whole cell, human MCF-7 whole cell, rat brain tissue, rat lung tissue, mouse brain tissue, mouse lung tissue. IHC: human lung cancer tissue, human mammary cancer tissue, human intestinal cancer tissue, human placenta tissue. ICC/IF: A431 cell, A431 cell. Flow: PC-3 cell, K562 cell, U251 cell.
Specifications
Specifications
| Antigen | CCT3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.01mg sodium azide |
| Gene | Cct3 |
| Gene Accession No. | P49368, P80318, Q6P502 |
| Gene Alias | AL024092; CCT3; CCTG; CCT-gamma; chaperonin containing TCP1 subunit 3; chaperonin containing Tcp1, subunit 3 (gamma); chaperonin subunit 3 (gamma); hTRiC5; Matricin; mTRiC-P5; OTTHUMP00000025735; PIG48; T-complex protein 1 subunit gamma; T-complex protein 1, gamma subunit; TCP1 (t-complex-1) ring complex, polypeptide 5; TCP-1-gamma; Tcp1-rs3; TRIC5; TriC-P5 |
| Gene Symbols | Cct3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.