Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 215241AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-3464


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The chaperonin containing TCP1 complex (CCT), also called the TCP1 ring complex, consists of 2 back-to-back rings, each containing 8 unique but homologous subunits, such as CCT4. CCT assists the folding of newly translated polypeptide substrates through multiple rounds of ATP-driven release and rebinding of partially folded intermediate forms. Substrates of CCT include the cytoskeletal proteins actin and tubulin, as well as alpha-transducin.
Specifications

Specifications

CCT4
Polyclonal
Unconjugated
CCT4
CCT DELTA, CCT4, Cctd, SRB, Stimulator of TAR RNA binding, TCP 1 delta
Rabbit
Antigen Affinity Chromatography
RUO
10575, 12464, 29374
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P50991, P80315, Q7TPB1
CCT4
CCT4 Fusion Protein Ag16128
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.