Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT8 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501565153
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5153 Supplier Bethyl Laboratories, Inc Supplier No. A303446A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Chicken, Bovine and Orangutan

TCP-1 theta is theta subunit of the CCT (chaperonin containing TCP1 complex) chaperonin molecule which is abundant in the eukaryotic cytosol and may be involved in the transport and assembly of the newly synthesized proteins. Molecular chaperones assist in the protein folding upon ATP hydrolysis and TCP-1 theta may play a role in the assembly of the BBsome, a complex that is involved in ciliogenesis regulating transport vesicles to the cilia. Further, TCP-1 theta is also involved in the folding of actin and tubulin. The intact CCT complex is composed of eight polypeptides in a double ring structure. CCT is important within cells in the folding of the VHL tumor suppressor protein.

Specifications

Antigen CCT8
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CCT8
Gene Accession No. P42932, P50990
Gene Alias C21orf112, Cctq, D21S246, PRED71, CCT-theta, T-complex protein 1 subunit theta, T-complex protein 1, theta subunit, TCP-1-theta, renal carcinoma antigen NY-REN-15
Gene Symbols Cct8
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10694, 12469
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.