Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCT8 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CCT8 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7842
![]() ![]() |
Proteintech Group Inc
122631AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MA1-82683 recognizes the theta polypeptide of the CCT chaperonin molecule complex. The intact CCT complex is composed of eight polypeptides in a double ring structure. CCT is important within cells in aiding the folding of proteins including actin, tubulin and the VHL tumor suppressor protein.Specifications
CCT8 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Zebrafish | |
P42932, P50990 | |
10694, 12469, 288305, 394037 | |
CCT8 Fusion Protein Ag2926 | |
Primary | |
-20°C | |
CCT8 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C21orf112, CCT theta, CCT8, Cctq, D21S246, KIAA0002, PRED71, TCP 1 theta | |
Cct8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title