Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT8L2 Monoclonal Antibody (OTI1E12), TrueMAB™, OriGene
SDP

Catalog No. 502539791
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9791 30 μL
50-167-5805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9791 Supplier OriGene Supplier No. TA505305S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCT8L2 Monoclonal antibody specifically detects CCT8L2 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen CCT8L2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CCT8L2
Gene Accession No. Q96SF2
Gene Alias CCT8L2; CESK1; chaperonin containing TCP1 subunit 8 like 2; chaperonin containing TCP1, subunit 8 (theta)-like 2; putative T-complex protein 1 subunit theta-like 2; T-complex protein 1 subunit theta-like 2
Host Species Mouse
Immunogen Full length human recombinant protein of human CCT8L2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150160
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.