Learn More
CCT8L2 Mouse anti-Human, Clone: OTI4B7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505337

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Possible molecular chaperone; assists the folding of proteins upon ATP hydrolysis.Specifications
CCT8L2 | |
Monoclonal | |
Unconjugated | |
CCT8L2 | |
CESK1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
150160 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96SF2 | |
CCT8L2 | |
Full length human recombit protein of human CCT8L2 produced in HEk293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.