Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCZ1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 221591AP150UL

Description
CG028 belongs to the UPF0550 family.Specifications
CCZ1 | |
Polyclonal | |
Unconjugated | |
CCZ1 | |
C7orf28A, C7orf28B, CCZ1, CGI 43, H_DJ1163J12.2, UPF0550 protein C7orf28 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231874, 51622 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P86791, Q8C1Y8 | |
CCZ1 | |
CCZ1 Fusion Protein Ag17804 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction