Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCZ1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989152
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9152 Supplier Bioss Supplier No. BS9543R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CG028 belongs to the UPF0550 family.

Specifications

Antigen CCZ1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCZ1
Gene Accession No. P86791
Gene Alias AU022870; BOS_22968; C25H7orf28A; C7orf28A; C7orf28B; CCZ1; CCZ1 homolog, vacuolar protein trafficking and biogenesis associated; CCZ1 vacuolar protein trafficking and biogenesis associated; CCZ1 vacuolar protein trafficking and biogenesis associated homolog; CCZ1 vacuolar protein trafficking and biogenesis associated homolog (S. cerevisiae); CCZ1A; CCZ1B; CG14980-like; CGI-43; H_DJ1163J12.2; H_NH0577018.2; hypothetical protein LOC393103; LOC100357280; Vacuolar fusion protein CCZ1 homolog; Vacuolar fusion protein CCZ1 homolog B; Vacuolar fusion protein CCZ1 homolog-like; zgc:55344
Gene Symbols CCZ1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCZ1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51622
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.