Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCZ1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102070 Supplier Invitrogen™ Supplier No. PA5102070
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CG028.

CCZ1B gene ontology (GO) annotations include vesicle-mediated transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCZ1B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CCZ1B
Gene Accession No. P86790
Gene Alias C7orf28A; C7orf28B; CCZ1; CCZ1 homolog B, vacuolar protein trafficking and biogenesis associated; CCZ1 homolog, vacuolar protein trafficking and biogenesis associated B; CCZ1 vacuolar protein trafficking and biogenesis associated B; CCZ1 vacuolar protein trafficking and biogenesis associated homolog B; CCZ1B; CGI-43; H_DJ1163J12.2; H_NH0577018.2; Vacuolar fusion protein CCZ1 homolog; Vacuolar fusion protein CCZ1 homolog B; Vacuolar fusion protein CCZ1 homolog-like
Gene Symbols CCZ1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCZ1B(Accession P86790), corresponding to amino acid residues N56-V106.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221960
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.