Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD-1000 Medium

Catalog No. p-7084482
Click to view available options
Quantity:
500 g
1 L
10 kg
20 L

A chemically defined (CD), animal origin-free (AOF) basal medium that is ideally suited for further hydrolysate or chemically defined supplementation

CD-1000 Medium can be used in various culture vessels in batch, fed-batch, and profusion processes for both research and manufacturing applications.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Form Liquid
Product Type Cell Culture Media
For Use With (Application) Various culture vessels in batch, fed-batch, and profusion processes for both research and manufacturing applications
Quantity 500 g
Source Animal origin-free, chemically defined
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.