Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CD 2314
GREENER_CHOICE

Catalog No. 382410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-241-0 10 mg
38-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-241-0 Supplier Tocris Bioscience™ Supplier No. 3824/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective RARβ agonist

Potent and selective RARβ receptor agonist (Kd values are 145 and >3760 nM for RARβ and RARα receptors respectively; no binding detected at RARγ). Inhibits growth of human HNSCC 22B, 183A and 22A cells (IC50 values are 3.0, 5.7 and 8.0 μM respectively).

Chemical Identifiers

CAS 170355-37-0
Molecular Formula C23H24O2S

Specifications

Quantity 10 mg
Formula Weight 364.5
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 364.503
Chemical Name or Material CD 2314
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.