Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CD 2665
GREENER_CHOICESDP

Catalog No. 380050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-005-0 50 mg
38-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-005-0 Supplier Tocris Bioscience™ Supplier No. 3800/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective RARβ/γ antagonist

Selective RARβγ antagonist (KD values are 110, 306 and > 1000 nM for RARγ, RARβ and RARα respectively). Blocks retinoic acid-induced apoptosis ex vivo.

Chemical Identifiers

CAS 170355-78-9
Molecular Formula C31H34O5

Specifications

Quantity 50 mg
Formula Weight 486.6
Purity >98%
Molecular Weight (g/mol) 486.608
Chemical Name or Material CD 2665
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.