Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CD 3254
GREENER_CHOICE

Catalog No. 330210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-0210 10 mg
33-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-0210 Supplier Tocris Bioscience™ Supplier No. 3302/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective RXRα agonist.

Selective RXRα agonist; exhibits no activity at RARα, RARβ or RARγ receptors.

Chemical Identifiers

CAS 196961-43-0
Molecular Formula C24H28O3

Specifications

Quantity 10 mg
Formula Weight 364.48
Purity >97%
Molecular Weight (g/mol) 364.485
Chemical Name or Material CD 3254
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.