Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD-M6PR Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018398 0.1 mg
NBP120167 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018398 Supplier Novus Biologicals Supplier No. NBP1201670.1MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD-M6PR Polyclonal specifically detects CD-M6PR in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD-M6PR
Applications Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with No Preservative
Gene Accession No. P24668
Gene Alias 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR
Gene Symbols M6PR
Host Species Rabbit
Immunogen A synthetic peptide from the internal region of mouse MPRD (Cation dependent mannose-6-phosphate receptor) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4074
Test Specificity MPRD.
Reconstitution Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability.
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.