Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD-M6PR Antibody (SR1712), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CD-M6PR |
---|---|
Clone | SR1712 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
CD-M6PR Monoclonal antibody specifically detects CD-M6PR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
CD-M6PR | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
A synthesized peptide derived from human CD-M6PR (Uniprot #: P20645) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1712 | |
Western Blot, Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Immunology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4074 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title