Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD OptiCHO™ Medium

Catalog No. 50591551
Encompass_Preferred
Click to view available options
Quantity:
1 L
6 x 1 L

CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO's legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advancedCHO CD EfficientFeed™ Liquid Nutrient Supplement Kit feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.

When to Use CD OptiCHO™ Medium

  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is 'finicky,' i.e. hard to grow or adapt

When to Consider Another Gibco™ Medium

Consider CD FortiCHO™ medium when:

  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

Consider CD CHO medium when:

  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

Consider CD DG44 medium when:

  • You use parental DG44 or DXB11 CHO cells (dhfr-)

Dry Media Formulations

Gibco™ liquid media are available in an AGT™ (Advanced Granulation Technology) dry media format that is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. AGT™ media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their liquid media counterparts.

Design Your Own Media Specific to Your Needs

Whether it's a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1 X
Form Liquid
Product Type CD OptiCHO Medium
Serum Level Serum-free
With Additives Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine, No Phenol Red
Product Line Gibco™
Quantity 1 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Bottle
Included Antibiotics No antibiotics
Species Hamster
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Suspension Cell Culture
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.