Learn More
Invitrogen™ CD1 Monoclonal Antibody (76-7-4), Biotin

Description
CD1 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 76-7-4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | CD1.1 |
| Gene Accession No. | Q9XS72 |
| Gene Alias | CD1; CD1 antigen; CD1.1; CD1A; CD1A antigen, a polypeptide; CD1a molecule; cd1a1; CD1A1 antigen; CD1c; cluster of differentiation 1 A; cortical thymocyte antigen CD1A; differentiation antigen CD1-alpha-3; epidermal dendritic cell marker CD1a; FCB6; HTA1; hTa1 thymocyte antigen; LOC101111409; R4; T6; T-cell surface antigen T6/Leu-6; T-cell surface glycoprotein CD1a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.