Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD1 Monoclonal Antibody (76-7-4), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528762
Description
CD1 Monoclonal Antibody for Flow
The CD1 is a family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to the plasma membrane and to recycling vesicles of the early endocytic system. Alternative splicing results in multiple transcript variants.
Specifications
CD1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.5 | |
Q9XS72 | |
CD1.1 | |
Fresh dd miniature swine thymocytes. | |
1 mL | |
Primary | |
Pig | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
76-7-4 | |
PE | |
CD1.1 | |
CD1; CD1 antigen; CD1.1; CD1A; CD1A antigen, a polypeptide; CD1a molecule; cd1a1; CD1A1 antigen; CD1c; cluster of differentiation 1 A; cortical thymocyte antigen CD1A; differentiation antigen CD1-alpha-3; epidermal dendritic cell marker CD1a; FCB6; HTA1; hTa1 thymocyte antigen; LOC101111409; R4; T6; T-cell surface antigen T6/Leu-6; T-cell surface glycoprotein CD1a | |
Mouse | |
Purified | |
RUO | |
396785 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction