Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD10 Monoclonal Antibody (eBioCB-CALLA (CB-CALLA)), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD10 |
---|---|
Clone | eBioCB-CALLA (CB-CALLA) |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H054T03B06A
![]() |
Invitrogen™
H054T03B06A |
100 Tests |
Each for $430.00
|
|
|||||
H054T02B06A
![]() |
Invitrogen™
H054T02B06A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD10 Monoclonal antibody specifically detects CD10 in Human samples. It is validated for Flow CytometrySpecifications
CD10 | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P08473 | |
4311 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
eBioCB-CALLA (CB-CALLA) | |
Flow Cytometry | |
NovaFluor Blue 610-70S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase | |
IgG2b κ | |
Affinity chromatography | |
Mme |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title