Learn More
Invitrogen™ CD10 Monoclonal Antibody (LT10), PE

Description
This antibody reacts with an extracellular epitope of CD10 (CALLA - Common acute lymphatic leukemia antigen), a 100 kDa type II integral membrane protein.
Specifications
Specifications
| Antigen | CD10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | LT10 |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | Mme |
| Gene Accession No. | P08473 |
| Gene Alias | 6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase |
| Gene Symbols | Mme |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.