Learn More
Invitrogen™ CD10 Polyclonal Antibody

Description
In sandwich ELISAs, less than 35% cross-reactivity with recombinant human (rh) Neprilysin is observed and less than 0.2% cross-reactivity with rhNeprilysin-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CD10 |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Mme |
| Gene Accession No. | P07861, P08473, Q61391 |
| Gene Alias | 6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase |
| Gene Symbols | Mme |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.