Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD100 Mouse anti-Human, Clone: 1C1A6, Proteintech
SDP

Catalog No. 501737000 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7000 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7000 Supplier Proteintech Group Inc Supplier No. 665821IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B-cell signaling by augmenting B-cell aggregation and survival and down-regulating CD23 expression. CD72 is a lymphocyte receptor for the Class IV semaphorin CD100, which is involved in regulating B cell signaling.

Specifications

Antigen CD100
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1C1A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SEMA4D
Gene Accession No. Q92854
Gene Alias A8, BB18, C9orf164, CD100, coll 4, GR3, M sema G, SEMA4D, SEMAJ, Semaphorin 4D
Gene Symbols SEMA4D
Host Species Mouse
Immunogen CD100 Fusion Protein Ag26461
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.