Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD100 (SEMA4D) Mouse anti-Human, Clone: A8, Invitrogen™

Catalog No. PIMA528146
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
PIMA528146 500 μg
1 options

Catalog No. PIMA528146

Supplier: Thermo Scientific MA528146

Mouse Monoclonal Antibody

This antibody recognizes human CD226, approximately a 65 kDa glycoprotein, also known as DNAM1 (DNAX accessory molecule-1). CD226 is broadly expressed on T-cells, NK cells, platelets, monocytes and a subset of B cells.

SEMA4D is a cell surface receptor for PLXN1B and PLXNB2 that plays an important role in cell-cell signaling. Promotes reorganization of the actin cytoskeleton and plays a role in axonal growth cone guidance in the developing central nervous system. Regulates dendrite and axon branching and morphogenesis. Promotes the migration of cerebellar granule cells and of endothelial cells. Plays a role in the immune system; induces B-cells to aggregate and improves their viability (in vitro). Promotes signaling via SRC and PTK2B/PYK2, which then mediates activation of phosphatidylinositol 3-kinase and of the AKT1 signaling cascade. Interaction with PLXNB1 mediates activation of RHOA. Diseases associated with SEMA4D include Hemorrhagic Fever With Renal Syndrome and Osteonecrosis Of The Jaw.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD100 (SEMA4D)
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SEMA4D
Gene Accession No. Q92854
Gene Symbols SEMA4D
Host Species Mouse
Immunogen Human PHA activated lymphocytes
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.