Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD100 (Sema4D) Mouse, anti-Mouse, R718, Clone: BMA-12, BD OptiBuild™

Catalog No. BDB752016 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752016 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752016 Supplier BD Biosciences Supplier No. 752016
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BMA-12 monoclonal antibody specifically binds to CD100 which is also known as, Semaphorin-4D (Sema4D), or Collapsin-4 (Coll-4). CD100 is a ∼150 kDa type I transmembrane glycoprotein that belongs to the class IV Semaphorin subfamily within the Ig gene superfamily. CD100 is expressed by cells from a broad range of tissues including the nervous system, where it serves in axon guidance and synapse formation, and the kidney. It is expressed by most hematopoietic cells including T cells which express CD100 more abundantly than B cells and dendritic cell subsets. Its expression is significantly upregulated upon activation of these cell types. CD100 can costimulate T cell responses. For B cells and dendritic cells, CD100 interaction with its ligands, CD72 or Plexin B1, induces cellular activation and promotes survival, respectively. CD100 is also known to interact with Plexin B2 on skin gamma delta T cells and contribute to the repair of tissue damage caused by inflammation. A biologically-active, soluble form of CD100 is generated by proteolytic cleavage of the cell surface form.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD100 (Sema4D)
Applications Flow Cytometry
Classification Monoclonal
Clone BMA-12
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.