Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD101 Monoclonal Antibody (Moushi101), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$194.00 - $375.00
Specifications
Antigen | CD101 |
---|---|
Clone | Moushi101 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M045T03B08A
![]() |
Invitrogen™
M045T03B08A |
100 μg |
Each for $375.00
|
|
|||||
M045T02B08A
![]() |
Invitrogen™
M045T02B08A |
25 μg |
Each for $194.00
|
|
|||||
Description
CD101 Monoclonal antibody specifically detects CD101 in Mouse samples. It is validated for Flow CytometrySpecifications
CD101 | |
0.1 mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
A8E0Y8 | |
630146 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Moushi101 | |
Flow Cytometry | |
NovaFluor Blue 660-120S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Cd101; CD101 antigen; CD101 molecule; cell surface glycoprotein V7; EWI immunoglobulin subfamily member EWI-101; EWI101; EWI-101; Glu-Trp-Ile EWI motif-containing protein 101; Gm1016; Gm734; IGSF2; Immunoglobulin superfamily member 2; immunoglobulin superfamily, member 2; leukocyte surface protein; RP11-27K13.2; V7 | |
IgG2a κ | |
Affinity chromatography | |
CD101 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title