Learn More
Description
Specifications
Specifications
| Antigen | CD101 |
| Applications | Single Cell 3' Sequencing |
| Classification | Monoclonal |
| Clone | Clone V7.1 |
| Conjugate | Oligo |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | IGSF2; Cell surface glycoprotein V7; V7; EWI-101; P126 |
| Host Species | Mouse |
| Immunogen | T cell clone CS1 |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.