Learn More
Description
The V7.1 monoclonal antibody specifically recognizes CD101 which is also known as P126 or Glu-Trp-Ile EWI motif-containing protein 101 (EWI-101). CD101 is encoded by IGSF2 (Immunoglobulin superfamily member 2) and belongs to the EWI (sharing a conserved glutamine-tryptophan-isoleucine motif) family within the Ig superfamily. CD101 is a ñ135-140 kDa type I transmembrane glycoprotein that has a short cytoplasmic domain with potential phosphorylation sites and an extracellular region with seven immunoglobulin-variable (IgV)-like domains. CD101 is expressed as homodimers by granulocytes, monocytes, dendritic cells, and some T cells. Its expression by T cells may be upregulated upon activation through the CD3 complex. CD101 may play a role in TCR/CD3-dependent T-cell activation.
Specifications
Specifications
| Antigen | CD101 |
| Classification | Monoclonal |
| Clone | V7.1 |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | IGSF2, Cell surface glycoprotein V7, V7, EWI-101, P126 |
| Host Species | Mouse |
| Immunogen | T cell clone CS1 |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.