Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD101 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980976
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0976 Supplier Bioss Supplier No. BS10727R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD101 also known as V7 antigen, a member of the Ig superfamily. CD101 is a disulfide linked homodimer of unknown function. Expression of CD101 is found on monocytes, granulocytes and dendritic cells (Langerhan-like cells HLA-DR, CD1a, CD1c). In addition expression on T lymphocytes is important for cell activation. CD101+ CD28+ cells are very responsive to CD28 signaling. In combination with anti-CD28 or suboptimal levels of anti-CD3, anti-CD101 can increase proliferation thereby suggesting an activating role. The monoclonal antibody BB27 has been shown to inhibit the T cell reactivity in allogeneic and antigen-specific mixed DC-T cell cultures. Recently it has been demonstrated that mouse CD101 is found on a subpopulation of regulatory T cells (CD4, CD25, Foxp3 positive) that have high suppressor activity. Expression of CD101 on human PBMCs shows staining of about 30% of the Foxp3 positive cells. Studies have not confirmed higher suppressor activity in the human CD101 population.

Specifications

Antigen CD101
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CD101
Gene Accession No. Q93033
Gene Alias CD101; CD101 antigen; CD101 molecule; Cell surface glycoprotein V7; EWI immunoglobulin subfamily member EWI-101; EWI101; EWI-101; Glu-Trp-Ile EWI motif-containing protein 101; Gm1016; Gm734; IGSF2; Immunoglobulin superfamily member 2; immunoglobulin superfamily, member 2; leukocyte surface protein; RP11-27K13.2; V7
Gene Symbols CD101
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CD101.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9398
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.