Learn More
Invitrogen™ CD102 (ICAM-2) Monoclonal Antibody (3C4), Biotin

Description
CD102 (ICAM-2) Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD102 (ICAM-2) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3C4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | ICAM2 |
| Gene Accession No. | P35330 |
| Gene Alias | CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.