Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD102 (ICAM-2) Monoclonal Antibody (3C4 (mIC2/4)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 53102182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-102-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-102-182 Supplier Invitrogen™ Supplier No. 53102182
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody 3C4 (mIC2/4) recognizes mouse CD102, also known as intercellular adhesion molecule-2 (ICAM-2). CD102, a member of the Immunoglobulin superfamily of receptors with 2 extracellular Ig domains, binds LFA (CD11a/CD18) and Mac-1 (CD11b/CD18), thereby mediating cellular interactions to lymphocytes. CD102 plays a role in NK cytotoxicity and migration, and lymphocyte recirculation. Expression of CD102 is found on most lymphocytes, monocytes and platelets. The 3C4 (mIC2/4) antibody blocks interaction of lymphocytes with antigen presenting cells. Applications Reported: This 3C4 (mIC2/4) antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applications Tested: This 3C4 (mIC2/4) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD102 (ICAM-2)
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 3C4 (mIC2/4)
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ICAM2
Gene Accession No. P35330
Gene Alias CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001
Gene Symbols ICAM2
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15896
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.