Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD102 (ICAM-2) Monoclonal Antibody (CBR-IC2/2), Biotin, eBioscience™
GREENER_CHOICE

Catalog No. 5017728
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-177-28 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-177-28 Supplier Invitrogen™ Supplier No. BMS109BT
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BMS109 does not cross react with mouse. Intercellular Adhesion Molecule-2 (ICAM-2) is a 60 kDa surface glycoprotein that promotes adhesion between immune cells and the vascular endothelium. ICAM-2 is a member of the Ig-superfamily with two immunoglobulin-like extracellular domains. ICAM-2 is much more closely related to the two N-terminal domains of ICAM-1 (34% identity) than either ICAM-1 or ICAM-2 is to other members of the Ig-superfamily, demonstrating the existence of a subfamily of Ig-like ligands that bind the same integrin receptor. ICAM-2 is broadly distributed on hematopoietic cells. On resting lymphocytes, the ICAM-2 expression is several-fold higher than that seen with ICAM-1, while monocytes express equivalent levels of ICAM-1 and ICAM-2. In contrast to ICAM-1, neutrophils stain negative for ICAM-2. Just little or no ICAM-2 staining can be demonstrated on other cell lines, the sole exception being resting vascular endothelial cells, which possess high levels of ICAM-2, and small clusters of cells in lymphoid tissue germinal centres. ICAM-2 expression is unaffected by a variety of inflammatory cytokines. Applications Tested: Flow Cytometry. Purity: >95%.

ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD102 (ICAM-2)
Applications Flow Cytometry
Classification Monoclonal
Clone CBR-IC2/2
Concentration 10 μL/Test
Conjugate Biotin
Formulation TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4
Gene ICAM2
Gene Accession No. P13598
Gene Alias CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001
Gene Symbols ICAM2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3384
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.