Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD102 Mouse anti-Human, Brilliant Violet 421, Clone: CBR-IC2/2, BD Optibuild™

Catalog No. BDB744536 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744536 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744536 Supplier BD Biosciences Supplier No. 744536
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CBR-IC2/2 monoclonal antibody specifically binds to CD102 which is also known as, intercellular adhesion molecule-2 (ICAM-2/ICAM2). CD102 is a type I transmembrane glycoprotein, member of the immunoglobulin supergene family, with an approximate molecular weight of 55-65 kDa. Its extracellular domain consists of two C2-type immunoglobulin-like subunits. The transmembrane region is 26 residues in size and the intracellular region also has 26 residues. CD102 is expressed on vascular endothelial cells, lymphocytes, monocytes, eosinophils, and platelets, but not on resting neutrophils. It is a ligand for the leukocyte integrin CD11a/CD18, or leukocyte function-associated antigen-1 (LFA-1), and there are reports of CD102 binding to leukocyte integrin CD11b/CD18 (Mac-1). Antibody CBR-IC2/2 blocks the binding of CD102 to leukocyte integrin CD11a/CD18. CD102 plays an important role in lymphocyte recirculation and in providing costimulatory signals in the immune response.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD102
Classification Monoclonal
Clone CBR-IC2/2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ICAM2, ICAM-2, Intercellular adhesion molecule 2
Host Species Mouse
Immunogen Human ICAM-2 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.