Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD102 Mouse, anti-Human, R718, Clone: CBR-IC2/2, BD OptiBuild™

Catalog No. BDB752240 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752240 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752240 Supplier BD Biosciences Supplier No. 752240
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CBR-IC2/2 monoclonal antibody specifically binds to CD102 which is also known as, intercellular adhesion molecule-2 (ICAM-2/ICAM2). CD102 is a type I transmembrane glycoprotein, member of the immunoglobulin supergene family, with an approximate molecular weight of 55-65 kDa. Its extracellular domain consists of two C2-type immunoglobulin-like subunits. The transmembrane region is 26 residues in size and the intracellular region also has 26 residues. CD102 is expressed on vascular endothelial cells, lymphocytes, monocytes, eosinophils, and platelets, but not on resting neutrophils. It is a ligand for the leukocyte integrin CD11a/CD18, or leukocyte function-associated antigen-1 (LFA-1), and there are reports of CD102 binding to leukocyte integrin CD11b/CD18 (Mac-1). Antibody CBR-IC2/2 blocks the binding of CD102 to leukocyte integrin CD11a/CD18. CD102 plays an important role in lymphocyte recirculation and in providing costimulatory signals in the immune response.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD102
Applications Flow Cytometry
Classification Monoclonal
Clone CBR-IC2/2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.