Learn More
CD102 Mouse, anti-Mouse, R718, Clone: 3C4(MIC2/4), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752096
Description
The 3C4 (mIC2/4) monoclonal antibody specifically binds to the mouse ICAM-2 (CD102) cell surface glycoprotein, a ligand for LFA-1. CD102 is constitutively expressed on endothelial cells, T and B lymphocytes, and alveolar walls. It is also expressed on a variety of leukocyte cell lines. CD102 does not appear to be involved in the development of hematopoietic cells. In a model for allergic asthma, endothelial CD102 mediates the transmigration of eosinophils (but not lymphocytes, monocytes, or macrophages) into the airway lumen. The 3C4 (mIC2/4) antibody blocks interactions between ICAM-2 and LFA-1.
Specifications
CD102 | |
Monoclonal | |
R718 | |
50 μg | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
3C4(MIC2/4) | |
Mouse | |
RUO | |
Mouse | |
Liquid |