Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD103 Hamster anti-Mouse , PE, Clone: 20000000, BD Biosciences™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 566844
Description
CD103 Monoclonal antibody specifically detects CD103 in Mouse samples. It is validated for Flow Cytometry
Specifications
CD103 | |
Monoclonal | |
PE | |
Itgae; integrin alpha-E; Integrin αIEL chain; αE; alpha-E1 | |
Mouse intestinal intraepithelial lymphocytes | |
RUO | |
Mouse | |
IgG2 κ |
Flow Cytometry | |
20000000 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction