Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD103 (Integrin alpha E) Monoclonal Antibody (B-Ly7), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$201.00 - $430.00
Specifications
Antigen | CD103 (Integrin alpha E) |
---|---|
Clone | B-Ly7 |
Concentration | 0.6 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H075T03Y03A
![]() |
Invitrogen™
H075T03Y03A |
100 Tests |
Each for $430.00
|
|
|||||
H075T02Y03A
![]() |
Invitrogen™
H075T02Y03A |
25 Tests |
Each for $201.00
|
|
|||||
Description
CD103 (Integrin alpha E) Monoclonal antibody specifically detects CD103 (Integrin alpha E) in Human samples. It is validated for Flow CytometrySpecifications
CD103 (Integrin alpha E) | |
0.6 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P38570 | |
3682 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
B-Ly7 | |
Flow Cytometry | |
NovaFluor Yellow 610 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
IgG1 κ | |
Affinity chromatography | |
Itgae |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title