Learn More
Invitrogen™ CD103 (Integrin alpha E) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117166
Description
Positive Samples: 293T, SW620, HeLa, Mouse lung, Mouse thymus Immunogen sequence: FNVDVARPWL TPKGGAPFVL SSLLHQDPST NQTWLLVTSP RTKRTPGPLH RCSLVQDEIL CHPVEHVPIP KGRHRGVTVV RSHHGVLICI QVLVRRPHSL SSELTGTCSL LGPDLRPQAQ ANFFDLENLL DPDARVDTGD CYSNKEGGGE DDVNTARQRR ALEKEEEEDK EEEEDEEEEE AG.
CD103 (ITGAE, Integrin alpha E) is molecule that is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. In particular, CD103 includes a 150kDa alpha chain and a 120kDa beta chain. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is part of the integrin family which are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. ITGAE encodes an I-domain-containing alpha integrin that undergoes post-translational cleavage in the extracellular domain, yielding disulfide-linked heavy and light chains. In combination with the beta 7 integrin, CD103 protein forms the E-cadherin binding integrin known as the human mucosal lymphocyte-1 antigen. CD103 is preferentially expressed in human intestinal intraepithelial lymphocytes (IEL), and in addition to a role in adhesion, it may serve as an accessory molecule for IEL activation. Diseases associated with CD103 dysfunction include hairy cell leukemia and splenic marginal zone lymphoma.
Specifications
CD103 (Integrin alpha E) | |
Polyclonal | |
Unconjugated | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Rabbit | |
Affinity chromatography | |
RUO | |
16407, 3682, 83577 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P38570, Q60677 | |
Itgae | |
Recombinant fusion protein containing a sequence corresponding to amino acids 19-200 of human ITGAE (NP_0021993). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.