Learn More
Invitrogen™ CD103 Monoclonal Antibody (OX-62)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA181573
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat OX-62 antibody, clone OX-62 recognizes the OX-62 antigen, also known as rat alpha E2 integrin, an 1150 amino acid, approximately 130 kDa single pass type I transmembrane protein expressed by intestinal dendritic cells, dendritic epidermal T cells, intraepithelial lymphocytes in the small intestine and by cells of dendritic morphology in lymphoid organs, at sites where gamma delta T cells are present.
CD103 (ITGAE, Integrin alpha E) is molecule that is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. In particular, CD103 includes a 150kDa alpha chain and a 120kDa beta chain. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is part of the integrin family which are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. ITGAE encodes an I-domain-containing alpha integrin that undergoes post-translational cleavage in the extracellular domain, yielding disulfide-linked heavy and light chains. In combination with the beta 7 integrin, CD103 protein forms the E-cadherin binding integrin known as the human mucosal lymphocyte-1 antigen. CD103 is preferentially expressed in human intestinal intraepithelial lymphocytes (IEL), and in addition to a role in adhesion, it may serve as an accessory molecule for IEL activation. Diseases associated with CD103 dysfunction include hairy cell leukemia and splenic marginal zone lymphoma.
Specifications
CD103 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
Itgae | |
Density gradient enriched PVG rat thoracic duct dendritic cells. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
OX-62 | |
Unconjugated | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Mouse | |
Protein A | |
RUO | |
83577 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.