Learn More
CD103 Mouse anti-Human, FITC, Clone: BER-ACT8, BD
Mouse Monoclonal Antibody
$128.00 - $262.00
Specifications
| Antigen | CD103 |
|---|---|
| Clone | BER-ACT8 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB561677
|
BD Biosciences
561677 |
25 Tests |
Each for $128.00
|
|
|||||
|
BDB550259
|
BD Biosciences
550259 |
100 Tests |
Each for $262.00
|
|
|||||
Description
The Ber-ACT8 monoclonal antibody specifically binds to the human mucosal lymphocyte antigen 1 (HML-1). This is a 175 kDa type I transmembrane glycoprotein, also known as integrin αE (ITGAE, Integrin alpha-E) and CD103. It is found on >90% of intestinal intraepithelial lymphocytes (iIEL) associated with integrin β7. CD103 is also expressed on lamina propria T lymphocytes in the intestine and on phytohemagglutinin-stimulated peripheral blood lymphocytes. It is rarely expressed on resting peripheral blood lymphocytes. It has been suggested that CD103 may have an accessory function for activation of iIEL. CD103 has been reported as a useful tool in hairy cell leukemia research.
Specifications
| CD103 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| BER-ACT8 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human MAPS16 T Cell Line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.