Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD104 (Integrin beta 4) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579541
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579541 Supplier Invitrogen™ Supplier No. PA579541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human PC-3 whole cell, human T-47D whole cell, human Hela whole cell, human Caco-2 whole cell. IHC: human placenta tissue, human intestinal cancer tissue. Flow: A431 cell.

Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD104 (Integrin beta 4)
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene ITGB4
Gene Accession No. P16144
Gene Alias AA407042; C230078O20; CD antigen CD104; CD104; CD104 antigen; GP150; integrin beta 4; integrin beta-4; integrin subunit beta 4; integrin, beta 4; Itgb4
Gene Symbols ITGB4
Host Species Rabbit
Immunogen E.coli-derived human Integrin beta 4 recombinant protein (Position: N28-A266).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3691
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.