Learn More
Invitrogen™ CD104 Monoclonal Antibody (422325), Alexa Fluor™ 488

Description
In Western blots, no cross-reactivity with recombinant human Integrin beta 1, beta 2, beta 3, beta 5, beta 6, beta 8, or recombinant mouse Integrin beta 4 is observed.
Specifications
Specifications
| Antigen | CD104 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 422325 |
| Conjugate | Alexa Fluor 488 |
| Formulation | Buffered saline solution with BSA and 0.09% sodium azide |
| Gene | ITGB4 |
| Gene Accession No. | P16144 |
| Gene Alias | AA407042; C230078O20; CD antigen CD104; CD104; CD104 antigen; GP150; integrin beta 4; integrin beta-4; integrin subunit beta 4; integrin, beta 4; Itgb4 |
| Gene Symbols | ITGB4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.