Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD104 Monoclonal Antibody (CL6973)
GREENER_CHOICE

Catalog No. PIMA535039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535039 Supplier Invitrogen™ Supplier No. MA535039
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: MKKVLVDNPK NRMLLIENLR ESQPYRYTVK ARNGAGWGPE REAIINLATQ PKRPMSIPII PDIPIVDAQS GEDYDSFLMY SDDVLRS Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.

Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD104
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL6973
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITGB4
Gene Accession No. P16144
Gene Alias AA407042; C230078O20; CD antigen CD104; CD104; CD104 antigen; GP150; integrin beta 4; integrin beta-4; integrin subunit beta 4; integrin, beta 4; Itgb4
Gene Symbols ITGB4
Host Species Mouse
Immunogen Recombinant protein fragment of Human ITGB4 (Product #RP-102714)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.