Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD104 Rat anti-Human, Brilliant Violet 510, Clone: 439-9B, BD Optibuild™

Catalog No. BDB744151 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744151 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744151 Supplier BD Biosciences Supplier No. 744151
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 439-9B monoclonal antibody specifically recognizes CD104, integrinβ4 chain, a 205 kDa transmembrane glycoprotein, which associates with CD49f (integrinα6 chain) to form theα6/β4 (CD49f/CD104) complex. It is expressed on epithelial cells, Schwann cells, and some tumor cells. The CD49f/CD104 complex is located in the hemidesmosomes of the epidermis, suggesting its role in epidermal cell-basement membrane adhesion. The clone 439-9B was clustered as CD104 at the fifth Human Leucocyte Differentiation Antigen International Workshop. It may be used for immunoprecipitation, immunoblotting and immunohistochemistry on frozen tissue sections.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD104
Classification Monoclonal
Clone 439-9B
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Integrin β4 chain, Integrin beta 4, ITGB4, GP150, TSP-180
Host Species Rat
Immunogen Human CD104 Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.