Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD105 (Endoglin) Monoclonal Antibody (MJ7/18), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544188
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4188 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4188 Supplier Leinco Technologies Inc Supplier No. C911100MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specificity: MJ7/18 activity is directed against mouse CD105 (endoglin). Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: =95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -70C. Avoid Repeated Freeze Thaw Cycles.

CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Mouse samples. It is validated for Affinity Purification, ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen CD105 (Endoglin)
Applications Affinity Purification, ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MJ7/18
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene ENG
Gene Accession No. Q63961
Gene Alias AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein
Host Species Rat
Immunogen Inflamed mouse skin
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13805
Target Species Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.