Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD105 (Endoglin) Mouse anti-Human, Alexa Fluor™ 647, Clone: SN6h (also known as SN6H; G4-2C2), BD Phrmingen™
Mouse Monoclonal Antibody
$104.50 - $262.00
Specifications
Antigen | CD105 (Endoglin) |
---|---|
Clone | SN6h (also known as SN6H; G4-2C2) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Description
CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Human samples. It is validated for Flow cytometrySpecifications
CD105 (Endoglin) | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Human | |
END; endoglin; HHT1; ORW1 | |
Endoglin from Human Acute Lymphoblastic Leukemia Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
SN6h (also known as SN6H; G4-2C2) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
2022 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title