Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD105 Monoclonal Antibody (MEM-229), Alexa Fluor™ 488
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA562182

Description
CD105 Monoclonal Antibody for ICC/IF, IHC (F), Flow
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Specifications
CD105 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P17813, P37176 | |
ENG | |
Recombinant Vaccinia virus containing the human CD105 (L-isoform) cDNA. | |
100 Tests | |
Primary | |
Human, Pig | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
MEM-229 | |
Alexa Fluor 488 | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Mouse | |
Size-exclusion Chromatography | |
RUO | |
2022, 397096 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction