Learn More
Description
This antibody recognizes an extracellular epitope of CD105 (Endoglin), a 90 kDa type I integral membrane homodimer glycoprotein expressed on vascular endothelial cells (small and large vessels), activated monocytes and tissue macrophages, stromal cells of certain tissues including bone marrow, pre-B lymphocytes in fetal marrow and erythroid precursors in fetal and adult bone marrow; it is also present on syncytiotrophoblast on placenta throughout pregnancy. Western Blot: non-reducing conditions; Immunohistochemistry (Frozen): acetone fixation.
Specifications
Specifications
| Antigen | CD105 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | MEM-229 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | ENG |
| Gene Accession No. | P17813, P37176 |
| Gene Alias | AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
